siRNA Details
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024432.2 | NP_077752.1 | 2, 3 | 320 | ||
GenBank mRNA | AF272895.1 | - | 2, 3 | 335 | ||
AK008942.1 | - | 2, 3 | 329 | |||
AK009117.1 | - | 1, 2 | 102 | |||
AK075705.1 | - | 2, 3 | 348 | |||
AK082510.1 | - | 359 | ||||
AK148170.1 | - | 288 | ||||
AK161273.1 | - | 2, 3 | 341 | |||
BC016412.1 | - | 2, 3 | 249 | 4215883 | ||
BC049963.1 | - | 2, 3 | 320 | 2655935 |
