siRNA Details
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024432.2 | NP_077752.1 | 9, 10 | 1118 | ||
GenBank mRNA | AF272895.1 | - | 9, 10 | 1133 | ||
AK008942.1 | - | 9, 10 | 1126 | |||
AK009117.1 | - | 8, 9 | 900 | |||
AK075705.1 | - | 7, 8 | 2182 | |||
AK082510.1 | - | 1157 | ||||
AK131187.1 | - | 3, 4 | 2890 | |||
AK148170.1 | - | 1086 | ||||
AK161273.1 | - | 9, 10 | 1139 | |||
AK188370.1 | - | 1, 2 | 81 | |||
BC016412.1 | - | 9, 10 | 1047 | 4215883 | ||
BC049963.1 | - | 9, 10 | 1118 | 2655935 |

More Information
Other Species Targeted

