siRNA Details
Gene Symbol: 2310030G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025865.2 | NP_080141.1 | 2 | 93 | ||
GenBank mRNA | AK007611.1 | - | 2 | 109 | ||
AK007929.1 | - | 2 | 82 | |||
AK009541.1 | - | 2 | 100 | |||
BC027409.1 | - | 2 | 93 | 4035256 |
