siRNA Details
Gene Symbol: Ccdc91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025911.2 | NP_080187.2 | 4 | 422 | ||
GenBank mRNA | AK007017.1 | - | 3 | 346 | ||
AK007917.1 | - | 4 | 423 | |||
BC006734.1 | - | 4 | 327 | 3156328 | ||
BC049073.1 | - | 4 | 391 | 5707404 |
