siRNA Details
Gene Symbol: Ccdc91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025911.2 | NP_080187.2 | 11 | 1227 | ||
GenBank mRNA | AK007917.1 | - | 11 | 1228 | ||
AK010099.1 | - | 1 | 29 | |||
BC006734.1 | - | 11 | 1132 | 3156328 | ||
BC024946.1 | - | 6 | 485 | 4010938 | ||
BC049073.1 | - | 11 | 1196 | 5707404 |
