siRNA Details
Gene Symbol: Dph7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026044.3 | NP_080320.1 | 6 | 748 | ||
GenBank mRNA | AK013297.1 | - | 6 | 728 | ||
AK078448.1 | - | 6 | 711 | |||
AK152000.1 | - | 5 | 661 | |||
AK153537.1 | - | 5 | 661 | |||
BC056962.1 | - | 4 | 1398 | |||
BC086486.1 | - | 6 | 750 |
