siRNA Details
Gene Symbol: Ssu72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026899.3 | NP_081175.2 | 1, 2 | 645 | ||
GenBank mRNA | AK004446.1 | - | 1, 2 | 335 | ||
AK005220.1 | - | 1, 2 | 336 | |||
AK019168.1 | - | 1, 2 | 101 | |||
AK148131.1 | - | 1, 2 | 409 | |||
BC016544.1 | - | 1, 2 | 325 | 3497878 |
