siRNA Details
Gene Symbol: Clec2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168223.1 | NP_001161695.1 | 6 | 588 | ||
NM_001168224.1 | NP_001161696.1 | 6 | 700 | |||
NM_027562.4 | NP_081838.1 | 7 | 762 | |||
GenBank mRNA | AB091386.1 | - | 4 | 394 | ||
AK014570.1 | - | 7 | 763 | |||
AK137753.1 | - | 6 | 698 | |||
AY137341.1 | - | 6 | 700 | |||
AY137342.1 | - | 6 | 588 | |||
BC117543.1 | - | 7 | 752 | 40105656 | ||
JF431014.1 | - | 5 | 540 | |||
JF792238.1 | - | 6 | 599 | |||
JF792239.1 | - | 6 | 650 |
