siRNA Details
Gene Symbol: Noxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027744.1 | NP_082020.1 | 7 | 922 | ||
GenBank mRNA | AK017096.1 | - | 7 | 1053 | ||
BC125331.1 | - | 7 | 922 | 40129846 | ||
BC125333.1 | - | 7 | 922 | 40129848 |
