siRNA Details
Gene Symbol: Tmem80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141950.1 | NP_001135422.1 | 4 | 370 | ||
NM_027797.3 | NP_082073.1 | 4 | 439 | |||
GenBank mRNA | AK017460.1 | - | 4 | 440 | ||
AK049405.1 | - | 1 | 393 | |||
AK134821.1 | - | 4 | 371 | |||
AK169083.1 | - | 4 | 329 | |||
BC022756.1 | - | 4 | 314 | 4922223 |
