siRNA Details
Gene Symbol: Cyb5r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028057.2 | NP_082333.1 | 9 | 927 | ||
GenBank mRNA | AK005159.1 | - | 9 | 928 | ||
AK159663.1 | - | 9 | 897 | |||
AK167178.1 | - | 9 | 921 | |||
BC016266.1 | - | 9 | 1723 | 4924006 | ||
BC024618.1 | - | 9 | 905 | 4235599 |
