siRNA Details
Gene Symbol: 1700030K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028170.2 | NP_082446.2 | 5 | 1430 | ||
GenBank mRNA | AK089918.1 | - | 5 | 1372 | ||
AK133815.1 | - | 5 | 1434 | |||
AK157359.1 | - | 4 | 641 | |||
AK172202.1 | - | 4 | 639 | |||
BC008558.1 | - | 5 | 1384 | 3592293 |
