siRNA Details
Gene Symbol: Usp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028846.5 | NP_083122.1 | 25 | 2852 | ||
GenBank mRNA | AF449715.1 | - | 25 | 2803 | ||
AK006800.1 | - | 8 | 786 | |||
AK029890.1 | - | 2748 | ||||
AK047242.1 | - | 25 | 2771 | |||
AK054279.1 | - | 25 | 2795 | |||
AK087412.1 | - | 25 | 2764 | |||
AK163663.1 | - | 25 | 2762 | |||
AK173083.1 | - | 25 | 2754 | |||
BC019735.1 | - | 11 | 1052 | |||
BC037199.1 | - | 19 | 2236 | |||
BC079674.1 | - | 2741 | 30531994 |
