siRNA Details
Gene Symbol: Spata9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029343.3 | NP_083619.1 | 2 | 177 | ||
GenBank mRNA | AK006554.1 | - | 2 | 178 | ||
AK016416.1 | - | 2 | 171 | |||
BC048558.1 | - | 2 | 157 | 6703288 |
