siRNA Details
Gene Symbol: Rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021430.2 | NP_067405.1 | 5, 6 | 1381 | ||
GenBank mRNA | AB041584.1 | - | 5, 6 | 1011 | ||
AK013469.1 | - | 2, 3 | 274 | |||
BC029965.1 | - | 5, 6 | 1059 | |||
BC051945.1 | - | 5, 6 | 1381 | 5716540 | ||
BC051946.1 | - | 5, 6 | 1311 | 6403995 | ||
BC058384.1 | - | 5, 6 | 1274 | 5718399 |
