siRNA Details
Gene Symbol: Mff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029409.2 | NP_083685.2 | 3 | 453 | ||
GenBank mRNA | AK017226.1 | - | 3 | 453 | ||
AK083056.1 | - | 3 | 438 | |||
AK139649.1 | - | 3 | 432 | |||
AK177568.1 | - | 1 | 67 | |||
AK178118.1 | - | 1 | 67 | |||
AK180892.1 | - | 1 | 100 | |||
AK181587.1 | - | 1 | 67 | |||
AK183556.1 | - | 1 | 67 | |||
AK184380.1 | - | 1 | 67 | |||
AK188151.1 | - | 1 | 50 | |||
AK199555.1 | - | 1 | 67 | |||
AK207673.1 | - | 1 | 67 | |||
AK207808.1 | - | 1 | 17 | |||
AK209807.1 | - | 1 | 67 | |||
AK210863.1 | - | 1 | 67 | |||
AK212284.1 | - | 1 | 67 | |||
AK212704.1 | - | 1 | 67 | |||
AK212777.1 | - | 1 | 67 | |||
AK213477.1 | - | 1 | 67 | |||
AK214594.1 | - | 1 | 36 | |||
AK214622.1 | - | 1 | 67 | |||
AK215829.1 | - | 1 | 12 | |||
AK216685.1 | - | 1 | 17 | |||
AK218828.1 | - | 1 | 67 | |||
AK218934.1 | - | 3 | 310 | |||
AK219490.1 | - | 1 | 67 | |||
BC016597.1 | - | 1 | 164 | 3489844 | ||
BC059229.1 | - | 3 | 453 | 6413193 |

More Information
Other Species Targeted

Gene Ontology Categories:
