siRNA Details
Gene Symbol: Mff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029409.2 | NP_083685.2 | 3 | 364 | ||
GenBank mRNA | AK017226.1 | - | 3 | 364 | ||
AK083056.1 | - | 3 | 349 | |||
AK139649.1 | - | 3 | 343 | |||
AK178872.1 | - | 3 | 221 | |||
AK211221.1 | - | 3 | 221 | |||
AK217444.1 | - | 3 | 221 | |||
AK218934.1 | - | 3 | 221 | |||
BC016597.1 | - | 1 | 75 | 3489844 | ||
BC059229.1 | - | 3 | 364 | 6413193 |
