siRNA Details
Gene Symbol: 1700018B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029597.1 | NP_083873.1 | 1, 2 | 243 | ||
GenBank mRNA | AK006079.1 | - | 1, 2 | 243 | ||
BC147528.1 | - | 1, 2 | 158 | 9056520 | ||
BC147542.1 | - | 1, 2 | 158 | 9056533 | ||
BC172077.1 | - | 1, 2 | 158 |
