siRNA Details
Gene Symbol: Zfp449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030139.4 | NP_084415.1 | 5 | 1641 | ||
GenBank mRNA | AK036596.1 | - | 5 | 1642 | ||
BC082594.1 | - | 4 | 1531 | 30642886 | ||
BC145936.1 | - | 4 | 1531 | 40131239 | ||
BC145938.1 | - | 4 | 1531 | 40131241 |
