siRNA Details
Gene Symbol: Pum2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160219.1 | NP_001153691.1 | 14 | 2408 | ||
NM_001160220.1 | NP_001153692.1 | 14 | 2408 | |||
NM_001160221.1 | NP_001153693.1 | 14 | 2223 | |||
NM_001160222.1 | NP_001153694.1 | 13 | 1986 | |||
NM_030723.2 | NP_109648.2 | 14 | 2332 | |||
GenBank mRNA | AF315590.1 | - | 14 | 2348 | ||
AK048119.1 | - | 8 | 1277 | |||
AK122225.1 | - | 15 | 2256 | |||
AK134185.1 | - | 14 | 2222 | |||
AK141682.1 | - | 14 | 2270 | |||
AK147617.1 | - | 15 | 2356 | |||
AK161357.1 | - | 3 | 579 | |||
AK163964.1 | - | 13 | 1961 | |||
AY027917.1 | - | 14 | 2248 | |||
BC041773.1 | - | 12 | 1960 | 3986207 |
