siRNA Details
Gene Symbol: Clec2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289706.1 | NP_001276635.1 | 7 | 753 | ||
NM_001289707.1 | NP_001276636.1 | 6 | 678 | |||
NM_001289708.1 | NP_001276637.1 | 5 | 595 | |||
NM_020257.2 | NP_064653.1 | 6 | 666 | |||
GenBank mRNA | AF121352.1 | - | 6 | 639 | ||
AF350411.1 | - | 6 | 643 | |||
AK041724.1 | - | 5 | 1381 | |||
AY137344.1 | - | 6 | 677 | |||
AY137345.1 | - | 6 | 665 | |||
AY256574.1 | - | 7 | 744 | |||
AY256575.1 | - | 6 | 667 | |||
AY256576.1 | - | 5 | 584 | |||
BC139192.1 | - | 6 | 639 | 8862214 | ||
BC139193.1 | - | 6 | 639 | 8862215 | ||
BC145574.1 | - | 6 | 651 | 9054140 | ||
HQ713440.1 | - | 5 | 573 |
