siRNA Details
Gene Symbol: Pcdhb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053132.3 | NP_444362.3 | 1 | 1340 | ||
GenBank mRNA | AF326300.1 | - | 1 | 1136 | ||
AK029340.1 | - | 1 | 1319 | |||
AK034205.1 | - | 1 | 1342 | |||
AK043979.1 | - | 1 | 1299 | |||
AK046463.1 | - | 1 | 1323 | |||
AY013789.1 | - | 1 | 1136 |
