siRNA Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028119.5 | NP_082395.2 | 7 | 1172 | ||
GenBank mRNA | AY027937.1 | - | 7 | 1090 | ||
BC028955.1 | - | 6 | 1356 | |||
BC058589.1 | - | 4 | 1544 | |||
BC080821.1 | - | 6 | 1508 |

More Information
Other Species Targeted

Gene Ontology Categories:
