siRNA Details
Gene Symbol: Dbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016974.3 | NP_058670.2 | 3 | 980 | ||
GenBank mRNA | AK140243.1 | - | 3 | 857 | ||
BC018323.1 | - | 3 | 943 | 4195116 | ||
BC064094.1 | - | 1 | 1209 | |||
BC084676.1 | - | 3 | 329 |
