siRNA Details
Gene Symbol: Sphk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172561.1 | NP_001166032.1 | 6 | 3243 | ||
NM_020011.5 | NP_064395.2 | 6 | 3552 | |||
NM_203280.3 | NP_975009.1 | 7 | 3314 | |||
GenBank mRNA | AK016616.1 | - | 3 | 2332 | ||
AK154321.1 | - | 6 | 2982 |
Gene Symbol: Dbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016974.3 | NP_058670.2 | 4 | 1172 | ||
GenBank mRNA | AK140243.1 | - | 4 | 1049 | ||
AK178288.1 | - | 2 | 169 | |||
AK213757.1 | - | 1 | 15 | |||
BC018323.1 | - | 4 | 1135 | 4195116 | ||
BC064094.1 | - | 2 | 1401 | |||
BC084676.1 | - | 4 | 521 | |||
BP770661.1 | - | 1 | 368 |
