siRNA Details
Gene Symbol: Egr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289925.1 | NP_001276854.1 | 3 | 356 | ||
NM_001289927.1 | NP_001276856.1 | 2 | 230 | |||
NM_018781.3 | NP_061251.1 | 2 | 303 | |||
GenBank mRNA | AB221656.1 | - | 2 | 303 | ||
AF132128.1 | - | 2 | 303 | |||
AK141603.1 | - | 1 | 307 | |||
BC100734.1 | - | 2 | 302 | |||
BC103568.1 | - | 3 | 354 | 40045218 | ||
BC103569.1 | - | 2 | 298 | |||
BC103570.1 | - | 3 | 355 | 40045221 | ||
S40835.1 | - | 2 | 211 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017086, XM_006252240, XM_008770818, XM_006252239, U12428, and others.
Gene Ontology Categories:
