siRNA Details
Gene Symbol: Pou3f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008901.1 | NP_032927.1 | 1 | 266 | ||
GenBank mRNA | AK158302.1 | - | 1 | 299 | ||
BC052012.1 | - | 1 | 314 | |||
BC086670.1 | - | 277 | ||||
BC138657.1 | - | 1 | 341 | 8861677 | ||
BC145919.1 | - | 1 | 341 | 40131222 | ||
X66603.1 | - | 309 |
