siRNA Details
Gene Symbol: Slc25a51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009949.3 | NP_001009949.3 | 3 | 927 | ||
GenBank mRNA | AK051139.1 | - | 924 | |||
AK146788.1 | - | 3 | 889 | |||
AK150362.1 | - | 3 | 885 | |||
AK151688.1 | - | 3 | 885 | |||
AK152171.1 | - | 3 | 885 | |||
AK156168.1 | - | 3 | 885 | |||
AK156342.1 | - | 3 | 886 | |||
AK164726.1 | - | 3 | 909 | |||
AK165253.1 | - | 3 | 923 | |||
BC089000.1 | - | 2 | 822 | 6418170 |
