siRNA Details
Gene Symbol: Slc25a51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009949.3 | NP_001009949.3 | 3 | 1086 | ||
GenBank mRNA | AK051139.1 | - | 1083 | |||
AK146788.1 | - | 3 | 1048 | |||
AK150362.1 | - | 3 | 1044 | |||
AK151688.1 | - | 3 | 1044 | |||
AK152171.1 | - | 3 | 1044 | |||
AK156168.1 | - | 3 | 1044 | |||
AK156342.1 | - | 3 | 1045 | |||
AK164726.1 | - | 3 | 1068 | |||
AK165253.1 | - | 3 | 1082 | |||
BC089000.1 | - | 2 | 981 | 6418170 |
