siRNA Details
Gene Symbol: Kctd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172747.2 | NP_766335.1 | 6 | 942 | ||
GenBank mRNA | AF534881.3 | - | 6 | 928 | ||
AK075722.1 | - | 6 | 943 | |||
BC079607.1 | - | 6 | 935 | 30359035 |
