siRNA Details
Gene Symbol: Olfr518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146306.1 | NP_666418.1 | 1 | 319 | ||
GenBank mRNA | BC104128.1 | - | 1 | 319 | ||
BC104129.1 | - | 1 | 319 | |||
BC115846.1 | - | 1 | 318 | |||
BC127222.1 | - | 1 | 319 | 40044607 | ||
BC127223.1 | - | 1 | 319 | 40044609 |
