siRNA Details
Gene Symbol: Olfr550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147104.2 | NP_667315.2 | 1 | 533 | ||
GenBank mRNA | BC116960.1 | - | 1 | 533 | 40126279 | |
BC116964.1 | - | 1 | 533 | 40126283 |
