siRNA Details
Gene Symbol: Dusp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176933.4 | NP_795907.1 | 2 | 595 | ||
GenBank mRNA | AK053746.1 | - | 2 | 592 | ||
AK080964.1 | - | 2 | 595 | |||
AK165991.1 | - | 2 | 911 | |||
BC118941.1 | - | 2 | 566 | 40092218 | ||
BC118942.1 | - | 2 | 566 | 40092219 |
