siRNA Details
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019648.3 | NP_062622.1 | 3 | 883 | ||
GenBank mRNA | AB003144.1 | - | 2 | 194 | ||
AF434712.1 | - | 2 | 273 | |||
AK076804.1 | - | 2 | 226 | |||
AK090323.1 | - | 2 | 214 | |||
AK147097.1 | - | 2 | 288 | |||
AK159650.1 | - | 2 | 298 | |||
AK163318.1 | - | 2 | 300 | |||
AK165301.1 | - | 3 | 885 | |||
AK165845.1 | - | 2 | 278 | |||
AK189664.1 | - | 1 | 12 | |||
BC002213.1 | - | 2 | 295 | 3490341 | ||
BC010263.1 | - | 2 | 268 | |||
BC092052.1 | - | 2 | 260 | 3670685 |
