siRNA Details
Gene Symbol: Nudt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291046.1 | NP_001277975.1 | 4 | 560 | ||
NM_019837.2 | NP_062811.1 | 4 | 552 | |||
GenBank mRNA | AF264064.1 | - | 4 | 551 | ||
AK079658.1 | - | 2 | 47 | |||
BC016534.1 | - | 5 | 458 | 2615904 | ||
BC046805.1 | - | 4 | 553 | 6829170 | ||
BU919402.1 | - | 4 | 384 |
