siRNA Details
Gene Symbol: Pdcd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019745.4 | NP_062719.2 | 4 | 488 | ||
GenBank mRNA | AF159368.1 | - | 4 | 391 | ||
AK005594.1 | - | 4 | 417 | |||
AK075926.1 | - | 4 | 436 | |||
AK077308.1 | - | 4 | 465 | |||
AK137198.1 | - | 4 | 458 | |||
AK149182.1 | - | 4 | 394 | |||
AK160234.1 | - | 4 | 438 | |||
AK168857.1 | - | 4 | 422 | |||
AK169689.1 | - | 4 | 422 | |||
BC019650.1 | - | 4 | 428 | 4022513 | ||
BC086778.1 | - | 4 | 407 | 5689088 |
