siRNA Details
Gene Symbol: Aknaos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005960.1 | - | 113 | |||
AK017104.1 | - | 659 | ||||
AK018846.1 | - | 111 |
Gene Symbol: Akna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170885.1 | - | 3560 |
