siRNA Details
Gene Symbol: Zcchc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175126.4 | NP_780335.1 | 1 | 976 | ||
GenBank mRNA | AK053859.1 | - | 1 | 713 | ||
BC055755.1 | - | 1 | 959 | 6412435 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zcchc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175126.4 | NP_780335.1 | 1 | 976 | ||
GenBank mRNA | AK053859.1 | - | 1 | 713 | ||
BC055755.1 | - | 1 | 959 | 6412435 |