siRNA Details
Gene Symbol: Bambi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026505.2 | NP_080781.1 | 3 | 677 | ||
GenBank mRNA | AK011305.1 | - | 3 | 700 | ||
AK017303.1 | - | 3 | 677 | |||
AK045192.1 | - | 3 | 677 | |||
AK143185.1 | - | 3 | 692 | |||
AK164632.1 | - | 3 | 680 | |||
BC019378.1 | - | 3 | 659 | 5009872 |
Gene Symbol: Bambi-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040009.1 | - | 1 | 640 |
