siRNA Details
Gene Symbol: Stoml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026942.3 | NP_081218.3 | 4 | 616 | ||
GenBank mRNA | AK007508.1 | - | 2 | 285 | ||
AK045820.1 | - | 4 | 588 | |||
AK148324.1 | - | 515 | ||||
BC017536.1 | - | 5 | 868 | |||
BC037074.1 | - | 4 | 546 | 5037636 |
