siRNA Details
Gene Symbol: Atxn7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028139.5 | NP_082415.2 | 3 | 325 | ||
GenBank mRNA | AK013145.1 | - | 3 | 232 | ||
BC017546.1 | - | 2 | 126 | |||
BC100598.1 | - | 2 | 137 | |||
BY019350.1 | - | 2 | 249 | |||
BY203850.1 | - | 3 | 197 |
