siRNA Details
Gene Symbol: Smc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301412.1 | NP_001288341.1 | 1 | 40 | ||
NM_008017.4 | NP_032043.3 | 1 | 40 | |||
GenBank mRNA | AK017747.1 | - | 1 | 19 | ||
BC094380.1 | - | 1 | 12 | 30543190 | ||
CJ049961.1 | - | 1 | 41 |
