siRNA Details
Gene Symbol: Pex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014598.1 | - | 2 | 649 | ||
AK040411.1 | - | 639 | ||||
AK042663.1 | - | 2 | 649 | |||
AK044186.1 | - | 2 | 601 | |||
AK087637.1 | - | 2 | 871 | |||
AK089485.1 | - | 2 | 639 | |||
AK145961.1 | - | 2 | 662 | |||
AK147925.1 | - | 2 | 641 | |||
BC019144.1 | - | 2 | 632 | 5050939 | ||
BC058694.1 | - | 2 | 622 | 6811976 |
