siRNA Details
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242363.2 | NP_001229292.1 | 13 | 3334 | ||
NM_001242364.2 | NP_001229293.1 | 12 | 3285 | |||
NM_001242365.2 | NP_001229294.1 | 12 | 2691 | |||
NM_199009.3 | NP_950174.1 | 12 | 3243 | |||
GenBank mRNA | AK014591.1 | - | 1 | 125 | ||
AK085350.1 | - | 12 | 3239 | |||
AK122552.1 | - | 11 | 2992 | |||
AK154173.1 | - | 13 | 3279 | |||
AK155273.1 | - | 13 | 3279 | |||
AK159716.1 | - | 12 | 2646 | |||
AW121372.1 | - | 1 | 224 | |||
BC018464.1 | - | 12 | 3202 | |||
BC019738.1 | - | 4 | 1117 | |||
BC021532.1 | - | 10 | 2612 | |||
BC043717.1 | - | 11 | 2759 | |||
BC063093.1 | - | 12 | 3200 | 6332038 |
