siRNA Details
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242363.2 | NP_001229292.1 | 12 | 2952 | ||
NM_001242364.2 | NP_001229293.1 | 11 | 2903 | |||
NM_001242365.2 | NP_001229294.1 | 11 | 2309 | |||
NM_199009.3 | NP_950174.1 | 11 | 2861 | |||
GenBank mRNA | AK085350.1 | - | 11 | 2857 | ||
AK122552.1 | - | 10 | 2610 | |||
AK154173.1 | - | 12 | 2896 | |||
AK155273.1 | - | 12 | 2897 | |||
AK169996.1 | - | 12 | 2934 | |||
BC018464.1 | - | 11 | 2820 | |||
BC019738.1 | - | 3 | 735 | |||
BC021532.1 | - | 9 | 2230 | |||
BC043717.1 | - | 10 | 2377 | |||
BC063093.1 | - | 11 | 2818 | 6332038 |
