siRNA Details
Gene Symbol: 1700013D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177502.1 | NP_001170973.1 | 2 | 245 | ||
GenBank mRNA | AK005943.1 | - | 2 | 58 | ||
BC120816.1 | - | 2 | 228 | 40129739 | ||
BC120818.1 | - | 2 | 228 | 40129741 |
