siRNA Details
Gene Symbol: Pcdhb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053133.1 | NP_444363.1 | 1 | 2248 | ||
GenBank mRNA | AY013790.1 | - | 1 | 2248 | ||
BC116268.1 | - | 1 | 2249 | |||
BC116269.1 | - | 1 | 2241 | |||
BC127160.1 | - | 1 | 2248 | 40094058 |
