siRNA Details
Gene Symbol: C78339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033192.2 | NP_001028364.1 | 4 | 1055 | ||
GenBank mRNA | AK090371.1 | - | 4 | 439 | ||
AK141282.1 | - | 4 | 1056 | |||
AK171039.1 | - | 5 | 956 | |||
AK218359.1 | - | 3 | 334 |
