siRNA Details
Gene Symbol: D10Bwg1070e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156477.1 | - | 1 | 605 | ||
AK158707.1 | - | 1 | 541 | |||
R75120.1 | - | 1 | 102 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D10Bwg1070e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156477.1 | - | 1 | 605 | ||
AK158707.1 | - | 1 | 541 | |||
R75120.1 | - | 1 | 102 |