siRNA Details
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ117594.1 | - | 1 | 236 | ||
DQ117595.1 | - | 1 | 236 | |||
DQ117596.1 | - | 1 | 236 | |||
DQ117597.1 | - | 1 | 236 | |||
DQ117598.1 | - | 1 | 236 | |||
DQ117599.1 | - | 1 | 236 | |||
DQ153215.1 | - | 1 | 236 |
